N-(4-Fluoro-2-methyl-6-nitrophenyl)acetamide ≥95%

Supplier: AOBChem USA

Warning

15331-1G 15331-25G 15331-5G
76936-970EA 79.57 USD
76936-970 76936-972 76936-974
N-(4-Fluoro-2-methyl-6-nitrophenyl)acetamide ≥95%
N-(4-Fluoro-2-methyl-6-nitrophenyl)acetamide
Formula: C₉H₉FN₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD11520129
CAS Number: 159020-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR