Ethyl 5-methylbenzo[b]thiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

151846-1G 151846-25G 151846-500MG 151846-5G
76936-956EA 164.44 USD
76936-956 76936-958 76936-960 76936-962
Ethyl 5-methylbenzo[b]thiophene-2-carboxylate ≥97%
Ethyl 5-methylbenzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₂O₂S
MW: 220.2874 g/mol
Storage Temperature: Ambient
CAS Number: 101219-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR