6-Bromo-3-butoxy-2-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15151-25G 15151-5G
76936-948EA 275.84 USD
76936-948 76936-950
6-Bromo-3-butoxy-2-fluorophenylboronic acid ≥97%
6-Bromo-3-butoxy-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BBrFO₃
MW: 290.922 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08276825
CAS Number: 1072951-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR