2-(Dimethylamino)-5-pyrimidineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: N,N-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine

15019-25G 15019-5G
76936-934EA 325.7 USD
76936-934 76936-936
2-(Dimethylamino)-5-pyrimidineboronic acid pinacol ester ≥97%
2-(Dimethylamino)-5-pyrimidineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₂₀BN₃O₂
MW: 249.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368238
CAS Number: 1032759-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR