Ethyl 2-(3-Cyanophenyl)-2,2-difluoroacetate ≥97%

Supplier: AOBChem USA

Warning

147872-1G 147872-25G 147872-500MG 147872-5G
76936-912EA 131.56 USD
76936-912 76936-914 76936-916 76936-918
Ethyl 2-(3-Cyanophenyl)-2,2-difluoroacetate ≥97%
Ethyl 2-(3-Cyanophenyl)-2,2-difluoroacetate
Formula: C₁₁H₉F₂NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD16756772
CAS Number: 1249788-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR