4,4,5,5-Tetramethyl-2-(5-phenylthiophen-2-yl)-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

14703-1G 14703-25G 14703-5G
76936-884EA 62.6 USD
76936-884 76936-886 76936-888
4,4,5,5-Tetramethyl-2-(5-phenylthiophen-2-yl)-1,3,2-dioxaborolane ≥97%
4,4,5,5-Tetramethyl-2-(5-phenylthiophen-2-yl)-1,3,2-dioxaborolane
Formula: C₁₆H₁₉BO₂S
Storage Temperature: Ambient
MDL Number: MFCD08063141
CAS Number: 459409-74-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR