5-Hydroxy-2-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

14392-1G 14392-25G 14392-500MG 14392-5G
76936-856EA 231.28 USD
76936-856 76936-858 76936-860 76936-862
5-Hydroxy-2-methylphenylboronic acid ≥97%
5-Hydroxy-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₉BO₃
Storage Temperature: Ambient
MDL Number: MFCD17011972
CAS Number: 1375216-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR