6-Bromo-4-methyl-3-pyridineboronic acid ≥95%

Supplier: AOBChem USA

Warning

13469-1G 13469-25G 13469-500MG 13469-5G
76936-770EA 292.81 USD
76936-770 76936-772 76936-774 76936-776
6-Bromo-4-methyl-3-pyridineboronic acid ≥95%
6-Bromo-4-methyl-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BBrNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD09037489
CAS Number: 1072945-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR