6-(4-Fluorophenyl)-3-pyridineboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2-(4-Fluorophenyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-(4-Fluorophenyl)pyridine-3-boronic acid pinacol ester
13451-1G
13451-25G
13451-500MG
13451-5G
76936-754EA
233.4
USD
76936-754
76936-756
76936-758
76936-760
6-(4-Fluorophenyl)-3-pyridineboronic acid pinacol ester ≥97%
6-(4-Fluorophenyl)-3-pyridineboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₇H₁₉BFNO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD09037494 CAS Number: 1073354-81-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...