2-(Methylthio)-5-pyridineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-(Methylsulphanyl)pyridine-3-boronic acid pinacol ester, 6-(Methylthio)pyridine-3-boronic acid pinacol ester

13447-1G 13447-25G 13447-500MG 13447-5G
76936-740EA 132.62 USD
76936-740 76936-742 76936-744 76936-746
2-(Methylthio)-5-pyridineboronic acid pinacol ester ≥97%
2-(Methylthio)-5-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₈BNO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD09037495
CAS Number: 849934-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR