6-Bromo-2-fluoro-3-iodophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

13385-25G 13385-5G
76936-728EA 381.93 USD
76936-728 76936-730
6-Bromo-2-fluoro-3-iodophenylboronic acid ≥97%
6-Bromo-2-fluoro-3-iodophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BBrFIO₂
Storage Temperature: Ambient
MDL Number: MFCD09038418
CAS Number: 1072951-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR