6-(4,4,5,5-Tetramethyl1,3,2-dioxaboralan-2-yl)-2,3-dihydroinden-1-one ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-inden-1-one

13354-1G 13354-25G 13354-500MG 13354-5G
76936-716EA 203.7 USD
76936-716 76936-718 76936-720 76936-722
6-(4,4,5,5-Tetramethyl1,3,2-dioxaboralan-2-yl)-2,3-dihydroinden-1-one ≥97%
6-(4,4,5,5-Tetramethyl1,3,2-dioxaboralan-2-yl)-2,3-dihydroinden-1-one

Precaución: for Research Use Only

Formula: C₁₅H₁₉BO₃
MW: 258.121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08706296
CAS Number: 915402-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR