1-Boc-indole-5-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate

13160-25G 13160-5G
76936-708EA 286.45 USD
76936-708 76936-710
1-Boc-indole-5-boronic acid pinacol ester ≥97%
1-Boc-indole-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₂₆BNO₄
MW: 343.2251 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663895
CAS Number: 777061-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR