8-(3-Chlorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%

Supplier: AOBChem USA

Warning

130031-1G 130031-25G 130031-500MG 130031-5G
76936-694EA 233.4 USD
76936-694 76936-696 76936-698 76936-700
8-(3-Chlorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol ≥97%
8-(3-Chlorophenyl)-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: for Research Use Only

Formula: C₁₄H₁₇ClO₃
MW: 268.7359 g/mol
Storage Temperature: Ambient
CAS Number: 1297474-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR