8-phenyl-1,4-dioxaspiro[4.5]decan-8-ol ≥97%

Supplier: AOBChem USA

Warning

129612-1G 129612-25G 129612-500MG 129612-5G
76936-662EA 324.64 USD
76936-662 76936-664 76936-666 76936-668
8-phenyl-1,4-dioxaspiro[4.5]decan-8-ol ≥97%
8-phenyl-1,4-dioxaspiro[4.5]decan-8-ol

Precaución: for Research Use Only

Formula: C₁₄H₁₈O₃
MW: 234.291 g/mol
Storage Temperature: Ambient
CAS Number: 94112-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR