Furan-2-yl(4-(trifluoromethyl)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

128892-1G 128892-25G 128892-500MG 128892-5G
76936-650EA 353.28 USD
76936-650 76936-652 76936-654 76936-656
Furan-2-yl(4-(trifluoromethyl)phenyl)methanol ≥97%
Furan-2-yl(4-(trifluoromethyl)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₂H₉F₃O₂
MW: 242.1938 g/mol
Storage Temperature: Ambient
CAS Number: 1245321-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR