5-Methoxycarbonylthiophene-2-boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: Thiophene-2-carboxylic acid methyl ester-5-boronic acid, (5-(Methoxycarbonyl)thiophen-2-yl)boronic acid

12717-1G 12717-25G 12717-500MG 12717-5G
76936-634EA 236.59 USD
76936-634 76936-636 76936-638 76936-640
5-Methoxycarbonylthiophene-2-boronic acid ≥97% research grade
5-Methoxycarbonylthiophene-2-boronic acid
Formula: C₆H₇BO₄S
MW: 185.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08443761
CAS Number: 876189-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR