Methyl(4-Propylphenyl)Sulfane ≥97%

Supplier: AOBChem USA

Warning

124647-1G 124647-25G 124647-500MG 124647-5G
76936-618EA 350.1 USD
76936-618 76936-620 76936-622 76936-624
Methyl(4-Propylphenyl)Sulfane ≥97%
Methyl(4-Propylphenyl)Sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₄S
Storage Temperature: Ambient
CAS Number: 909785-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR