7-Bromo-4-methoxyindole ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 7-Bromo-4-methoxy-1H-indole

124403-1G 124403-25G 124403-500MG 124403-5G
76936-598EA 210.06 USD
76936-598 76936-600 76936-602 76936-604
7-Bromo-4-methoxyindole ≥97% research grade
7-Bromo-4-methoxyindole
Formula: C₉H₈BrNO
MW: 226.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04037872
CAS Number: 81224-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR