Potassium-2-formylphenyltrifluoroborate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Potassium trifluoro(2-formylphenyl)borate, Potassium 2-formylphenyltrifluoroborate

12418-25G 12418-5G
76936-582EA 198.39 USD
76936-582 76936-584
Potassium-2-formylphenyltrifluoroborate ≥95%
Potassium 2-formylphenyltrifluoroborate

Precaución: for Research Use Only

Formula: C₇H₅BF₃KO
MW: 212.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02093979
CAS Number: 192863-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR