Potassium (2,4-dimethylphenyl)trifluoroborate ≥95% research grade

Supplier: AOBChem USA

Warning

12415-25G 12415-5G
76936-576EA 310.85 USD
76936-576 76936-578
Potassium (2,4-dimethylphenyl)trifluoroborate ≥95% research grade
Potassium (2,4-dimethylphenyl)trifluoroborate
Formula: C₈H₉BF₃K
MW: 212.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368353
CAS Number: 850245-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR