3-Quinolineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Quinoline-3-boronic acid pinacolate, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline, Quinoline-3-boronic acid Pinacol ester, 4,4,5,5-Tetramethyl-2-(3-quinolyl)-1,3,2-dioxaborolane

12301-25G 12301-5G
76936-524EA 268.41 USD
76936-524 76936-526
3-Quinolineboronic acid pinacol ester ≥97%
3-Quinolineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₁₈BNO₂
MW: 255.1199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05155218
CAS Number: 171364-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR