5-Isoquinolineboronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
12123-25G
12123-5G
76936-488EA
463.62
USD
76936-488
76936-490
5-Isoquinolineboronic acid ≥97%
5-Isoquinolineboronic acid
Precaución: for Research Use Only
Formula:
C₉H₈BNO₂ MW: 172.98 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03839356 CAS Number: 371766-08-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...