Ethyl-4-bromo-2-hydroxybenzoate ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 4-bromo-2-hydroxybenzoate

115937-1G 115937-25G 115937-500MG 115937-5G
76936-448EA 175.05 USD
76936-448 76936-450 76936-452 76936-454
Ethyl-4-bromo-2-hydroxybenzoate ≥97% research grade
Ethyl-4-bromo-2-hydroxybenzoate
Formula: C₉H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD10566803
CAS Number: 314240-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR