6-Bromo-2-chloro-3-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

108190-1G 108190-25G 108190-500MG 108190-5G
76936-416EA 379.81 USD
76936-416 76936-418 76936-420 76936-422
6-Bromo-2-chloro-3-methoxypyridine ≥97%
6-Bromo-2-chloro-3-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClNO
MW: 222.467 g/mol
Storage Temperature: Ambient
CAS Number: 850220-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR