6-Bromo-2-pyridineboronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Bromo-2-pyridylboronic acid Pinacol ester, 6-Bromopyridine-2-boronic acid pinacol ester, 2-(6-Bromo-2-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

10440-25G 10440-5G
76936-384EA 230.22 USD
76936-384 76936-386
6-Bromo-2-pyridineboronic acid pinacol ester ≥95%
6-Bromo-2-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₅BBrNO₂
Storage Temperature: Ambient
MDL Number: MFCD06796177
CAS Number: 651358-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR