Methyl 3-amino-7-fluorobenzo[b]thiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

104039-1G 104039-25G 104039-500MG 104039-5G
76936-372EA 225.98 USD
76936-372 76936-374 76936-376 76936-378
Methyl 3-amino-7-fluorobenzo[b]thiophene-2-carboxylate ≥97%
Methyl 3-amino-7-fluorobenzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈FNO₂S
MW: 225.239 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29158206
CAS Number: 2090172-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR