Methyl-3-amino-6-bromobenzo[b]thiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

104037-1G 104037-25G 104037-5G
76936-364EA 70.02 USD
76936-364 76936-366 76936-368
Methyl-3-amino-6-bromobenzo[b]thiophene-2-carboxylate ≥97%
Methyl 3-amino-6-bromobenzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO₂S
Storage Temperature: Ambient
MDL Number: MFCD09865017
CAS Number: 1017782-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR