5-Pyrimidineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine, 2-(5-Pyrimidyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, Pyrimidine-5-boronic acid pinacol ester, Pyrimidyl-5-boronic acid pinacol ester

10250-25G 10250-5G
76936-344EA 187.78 USD
76936-344 76936-346
5-Pyrimidineboronic acid pinacol ester ≥97%
5-Pyrimidineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₀H₁₅BN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD05155223
CAS Number: 321724-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR