5-Methoxypyridine-3-boronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 5-methoxypyridin-3-yl boronic acid, (5-Methoxypyridin-3-yl)boronic acid, 5-Methoxy-3-pyridineboronic acid

10236-25G 10236-5G
76936-338EA 332.07 USD
76936-338 76936-340
5-Methoxypyridine-3-boronic acid ≥95%
5-Methoxypyridine-3-boronic acid

Precaución: for Research Use Only

Formula: C₆H₈BNO₃
Storage Temperature: Ambient
MDL Number: MFCD03425977
CAS Number: 850991-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR