5-Bromo-4-(difluoromethoxy)benzo[d][1,3]dioxole ≥97%

Supplier: AOBChem USA

Warning

99256-1G 99256-25G 99256-500MG 99256-5G
76936-148EA 412.7 USD
76936-148 76936-150 76936-152 76936-154
5-Bromo-4-(difluoromethoxy)benzo[d][1,3]dioxole ≥97%
5-Bromo-4-(difluoromethoxy)benzo[d][1,3]dioxole

Precaución: for Research Use Only

Formula: C₈H₅BrF₂O₃
Storage Temperature: Ambient
CAS Number: 2921834-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR