4-Bromo-5-chloro-2,3-dimethoxypyridine ≥97%

Supplier: AOBChem USA

Warning

95115-1G 95115-25G 95115-500MG 95115-5G
76936-132EA 270.53 USD
76936-132 76936-134 76936-136 76936-138
4-Bromo-5-chloro-2,3-dimethoxypyridine ≥97%
4-Bromo-5-chloro-2,3-dimethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₇BrClNO₂
Storage Temperature: Ambient
CAS Number: 1881289-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR