4-Chloro-3-fluoro-2-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

94128-1G 94128-500MG 94128-5G 94128-25G
76936-106EA 1464.05 USD
76936-106 76936-108 76936-110 77636-258
4-Chloro-3-fluoro-2-(methylthio)phenol ≥97%
4-Chloro-3-fluoro-2-(methylthio)phenol

Precaución: for Research Use Only

Formula: C₇H₆ClFOS
MW: 192.638 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34773896
CAS Number: 2707341-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR