5-Bromo-2-chloro-3-ethoxypyridine ≥97%

Supplier: AOBChem USA

Warning

91299-1G 91299-25G 91299-500MG 91299-5G
76936-092EA 281.14 USD
76936-092 76936-094 76936-096 76936-098
5-Bromo-2-chloro-3-ethoxypyridine ≥97%
5-Bromo-2-chloro-3-ethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₇BrClNO
MW: 236.494 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19688665
CAS Number: 1241752-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR