5-Bromo-2-fluoro-4-iodobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

90140-1G 90140-25G 90140-500MG 90140-5G
76936-082EA 298.12 USD
76936-082 76936-084 76936-086 76936-088
5-Bromo-2-fluoro-4-iodobenzoic acid ≥97%
5-Bromo-2-fluoro-4-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrFIO₂
MW: 344.904 g/mol
Storage Temperature: Ambient
CAS Number: 1807757-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR