4-Fluoro-2-iodo-6-methylaniline ≥97%

Supplier: AOBChem USA

Warning

81840-25G 81840-5G
76936-024EA 453.01 USD
76936-024 76936-026
4-Fluoro-2-iodo-6-methylaniline ≥97%
4-Fluoro-2-iodo-6-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇FIN
MW: 251.04 g/mol
Storage Temperature: Ambient
CAS Number: 1260885-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR