4'-Methyl-[1,1'-biphenyl]-4-amine ≥97%

Supplier: AOBChem USA

Warning

80573-1G 80573-25G 80573-5G
76936-012EA 78.51 USD
76936-012 76936-014 76936-016
4'-Methyl-[1,1'-biphenyl]-4-amine ≥97%
4'-Methyl-[1,1'-biphenyl]-4-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₃N
MW: 183.249 g/mol
Storage Temperature: Ambient
CAS Number: 1204-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR