5-Bromo-N,N,6-trimethylpyridin-2-amine ≥97%

Supplier: AOBChem USA

Warning

80059-1G 80059-25G 80059-500MG 80059-5G
76935-996EA 205.82 USD
76935-996 76935-998 76936-000 76936-002
5-Bromo-N,N,6-trimethylpyridin-2-amine ≥97%
5-Bromo-N,N,6-trimethylpyridin-2-amine

Precaución: for Research Use Only

Formula: C₈H₁₁BrN₂
MW: 215.09 g/mol
Storage Temperature: Ambient
CAS Number: 910054-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR