4-Methoxy-3,5-dimethylphenol ≥97%

Supplier: AOBChem USA

Warning

78642-1G 78642-25G 78642-500MG 78642-5G
76935-980EA 260.99 USD
76935-980 76935-982 76935-984 76935-986
4-Methoxy-3,5-dimethylphenol ≥97%
4-Methoxy-3,5-dimethylphenol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18432122
CAS Number: 4962-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR