4-Chloro-1-methyl-1H-pyrazole ≥97%
Supplier: AOBChem USA
|
Warning
|
78187-25G
78187-5G
76935-956EA
381.93
USD
76935-956
76935-958
4-Chloro-1-methyl-1H-pyrazole ≥97%
4-Chloro-1-methyl-1H-pyrazole
Formula:
C₄H₅ClN₂ Storage Temperature: Ambient |
MDL Number:
MFCD08275463 CAS Number: 35852-81-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...