5-Bromo-3-chloro-2-isopropoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

76297-1G 76297-25G 76297-500MG 76297-5G
76935-940EA 233.4 USD
76935-940 76935-942 76935-944 76935-946
5-Bromo-3-chloro-2-isopropoxy-1,1'-biphenyl ≥97%
5-Bromo-3-chloro-2-isopropoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₄BrClO
MW: 325.628 g/mol
Storage Temperature: Ambient
CAS Number: 2918836-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR