5-Bromo-2-fluoro-3-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

75675-1G 75675-25G 75675-500MG 75675-5G
76935-928EA 338.43 USD
76935-928 76935-930 76935-932 76935-934
5-Bromo-2-fluoro-3-(trifluoromethyl)phenol ≥97%
5-Bromo-2-fluoro-3-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₃BrF₄O
MW: 258.9957 g/mol
Storage Temperature: Ambient
CAS Number: 1807191-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR