4-Bromo-3-formylbenzonitrile ≥97%

Supplier: AOBChem USA

Warning

75101-1G 75101-25G 75101-5G
76935-902EA 62.6 USD
76935-902 76935-904 76935-906
4-Bromo-3-formylbenzonitrile ≥97%
4-Bromo-3-formylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₄BrNO
Storage Temperature: Refrigerator
MDL Number: MFCD11858309
CAS Number: 89003-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR