4-Fluoro-2-methyl-1-(methylsulfonyl)benzene ≥97% research grade

Supplier: AOBChem USA

Warning

74473-1G 74473-25G 74473-5G
76935-890EA 84.88 USD
76935-890 76935-892 76935-894
4-Fluoro-2-methyl-1-(methylsulfonyl)benzene ≥97% research grade
4-Fluoro-2-methyl-1-(methylsulfonyl)benzene
Formula: C₈H₉FO₂S
Storage Temperature: Ambient
MDL Number: MFCD04037935
CAS Number: 828270-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR