4-Hydroxy-[1,1'-biphenyl]-3-carboxylic acid ≥97% research grade

Supplier: AOBChem USA

Warning

74200-1G 74200-25G 74200-500MG 74200-5G
76935-874EA 205.82 USD
76935-874 76935-876 76935-878 76935-880
4-Hydroxy-[1,1'-biphenyl]-3-carboxylic acid ≥97% research grade
4-Hydroxy-[1,1'-biphenyl]-3-carboxylic acid
Formula: C₁₃H₁₀O₃
MW: 214.21 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00272245
CAS Number: 323-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR