4-Bromo-3-chloro-5-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

73331-1G 73331-500MG 73331-5G
76935-862EA 575.01 USD
76935-862 76935-864 76935-866
4-Bromo-3-chloro-5-fluorobenzoic acid ≥97%
4-Bromo-3-chloro-5-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrClFO₂
MW: 253.453 g/mol
Storage Temperature: Ambient
CAS Number: 1263274-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR