4-Chloro-6-methoxypyridine-3-carboxylic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-6-methoxynicotinic acid

72918-1G 72918-25G 72918-500MG 72918-5G
76935-846EA 292.81 USD
76935-846 76935-848 76935-850 76935-852
4-Chloro-6-methoxypyridine-3-carboxylic acid ≥97% research grade
4-Chloro-6-methoxypyridine-3-carboxylic acid
Formula: C₇H₆ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD13188727
CAS Number: 1060806-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR