4-Ethoxy-2-fluorobenzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-Ethoxy-2-Fluorobenzoic acid

72487-1G 72487-25G 72487-500MG 72487-5G
76935-830EA 281.14 USD
76935-830 76935-832 76935-834 76935-836
4-Ethoxy-2-fluorobenzoic acid ≥97% research grade
4-Ethoxy-2-fluorobenzoic acid
Formula: C₉H₉FO₃
Storage Temperature: Refrigerator
MDL Number: MFCD13196593
CAS Number: 1206593-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR