5-Ethyl-2,4-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

69814-1G 69814-500MG 69814-5G
76935-816EA 1331.43 USD
76935-816 76935-818 76935-820
5-Ethyl-2,4-difluorobenzaldehyde ≥97%
5-Ethyl-2,4-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈F₂O
MW: 170.156 g/mol
Storage Temperature: Ambient
CAS Number: 1783796-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR