4-Bromo-3-fluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

68968-1G 68968-500MG 68968-5G
76935-804EA 530.45 USD
76935-804 76935-806 76935-808
4-Bromo-3-fluoro-1,1'-biphenyl ≥97%
4-Bromo-3-fluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₈BrF
Storage Temperature: Ambient
CAS Number: 150805-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR